SpectraMax® Plus 384 Absorbance Plate Reader
Leverandør: MOLECULAR DEVICES
0310-3258
MLDV0310-3258EA 0 NOK
MLDV0310-3258
SpectraMax® Plus 384 Absorbance Plate Reader
Mikroplatelesere
This UV-visible absorbance microplate reader provides ultrafast, full spectral range detection for cuvettes, 96-well, and 384-well microplates, with the only temperature-independent method for pathlength correction.
  • Temperature control
  • Shaking capabilities
  • Multichannel optical design
  • Eliminates error
Temperature control and shaking come standard.
The patented multichannel optical design of the SpectraMax® Plus384 Absorbance Microplate Reader mimics a dual beam spectrophotometer, measuring each sample in the plate directly, eliminating measurementMore Product Information