Advanced Solutions BioAssemblyBot 400 Bioprinter
Leverandør: MOLECULAR DEVICES
BAB400
MLDVBAB400EA 0 NOK
MLDVBAB400
Advanced Solutions BioAssemblyBot 400 Bioprinter
Printere Bioprinters
Automated bioprinting solution for high-content screening with the BAB400 bioprinter for optimised 3D tissue and organoid workflow.
  • Develop new drug targets
  • Improved viability of 3D cell cultures
  • Enhanced flexibility with integrated workflows
Consistently dispense, place, and print organoids for use in drug discovery with higher throughput vs. manual workflows. Test drug toxicity or efficacy on 3D models that are representative of native human tissues.
The reliability and reproducibility of BAB allows for consistency in maintaining improvedMore Product Information