CloneDetect
Leverandør: MOLECULAR DEVICES
K8205
MLDVK8205EA 0 NOK
MLDVK8205
CloneDetect
Assay Cellulær analyse
Screen and select high value mammalian clones with label-free, in situ detection of IgG secretion with the ClonePix system, and other cell analysers with a green fluorescent channel.
  • Screen and select high-value mammalian clones with label-free, in situ detection of IgG secretion
  • A choice of detection agents specific for secreted mouse, rat or human IgGs
  • Sterile, virus and mycoplasma free
  • Optimised for ClonePix system, and compatible with cell analysers with a green fluorescent channel
A cell-culture validated, sterile and azide-free antibody reagent for detecting IgG in mammalian cell clone screening.