IMAP kinase, phosphatase, and phosphodiesterase assays, molecular devices
Leverandør: MOLECULAR DEVICES
R8160
MLDVR8160EA 0 NOK
MLDVR8160
IMAP kinase, phosphatase, and phosphodiesterase assays, molecular devices
Assay Enzymanalyser
IMAP® technology provides a homogeneous assay applicable to a wide variety of kinases without regard for substrate peptide sequence.
  • Complete assay system
  • Robust fluorescence signal
  • Homogeneous
  • Available in both FP and TR-FRET detection modes
IMAP provides a complete assay system for screening kinases, phosphatases, and phosphodiesterases
The assay is a simple "mix-and-read" procedure that allows accurate determination of kinase, phosphatase, and phosphodiesterase (PDE) activity.
IMAP Binding Reagent and Binding Buffers for 8000 data points in a standard 384 well format.