Abrasive sheet, sand paper, Fre-Cut™ 618
Leverandør: 3M
02636
MMMA02636EA 1600 NOK
MMMA02636
Abrasive sheet, sand paper, Fre-Cut™ 618
Papirservietter Rengjøringssett og svamper
These flexible ‘A’ weight high performance silicon carbide sheets are ideal for finishing paints and lacquers, wood and plastics. For hand sanding or random orbital sanding on furniture, ornaments, frames, fillers, mouldings, profiles.
  • Sharp silicon carbide mineral gives an aggressive cut with fine finish
  • Higher loading resistance means extended product life
Grid grade: P500
Mineral: Silicon carbide
Backing: A-weight paper
Coating: Open
Bond: Resin