Value masking tape 101E
Leverandør: 3M
101E-48MM 101E-18MM 101E-24MM 101E-30MM
MMMA101E-48MMEA 97 NOK
MMMA101E-48MM MMMA101E-18MM MMMA101E-24MM MMMA101E-30MM
Value masking tape 101E
Tape
This is a smooth, multi-purpose crepe paper tape for room temperature, indoor, light-duty applications. It is ideal for non-critical paint masking, but also bundling, sealing, attaching, and other applications.
  • Good initial tack to a wide variety of surfaces in different applications
  • Designed for light-duty applications
  • Operating temperature range up to 30 °C
  • Easy to write on for labelling
The cost-effective general-purpose value masking tape is a chamois-colored, 0,12 mm thick adhesive tape with rubber adhesive for general applications (temporaryMore Product Information