DBI-SALA® ExoFit™ XE200 Comfort Rescue Safety Harnesses
Leverandør: 3M
1112733 1112734
MMMA1112733EA 6750 NOK
MMMA1112733 MMMA1112734
DBI-SALA® ExoFit™ XE200 Comfort Rescue Safety Harnesses
Fallvern Fallsele
Cross chest buckle assembly for central front fall arrest attachment point - optimised position on body. Revolving torso adjusters (patented) for quick and easy harness fit and adjustment. Auto-locking quick connect buckles secure contact with webbing, reducing the need to readjust.
  • Integrated pSRL tunnel lets you quickly and easily connect and disconnect personal SRLs
  • Equipped with auto-resetting and re-positionable lanyard keepers for fast and easy parking of snap hooks and carabiners
  • Suspension trauma relief straps with low-profile design included
  • 3M™