Scotch® 130C Linerless Rubber Splicing Tape
Leverandør: 3M
130C
MMMA130CEA 18375 NOK
MMMA130C
Scotch® 130C Linerless Rubber Splicing Tape
Tape
Scotch® Liner less rubber splicing tape 130C is a highly conformable, liner less, ethylene rubber (EPR), high voltage insulating tape. This self-bonding tape has excellent physical and electrical properties, which provide immediate moisture seals as well as void-free build-ups.
  • Highly conformable, rubber-resin adhesive
  • Self-fusing, ethylene propylene, rubber backing provides UV resistance
  • Designed to insulate splices and terminate cables whose overload temperatures can reach 130 °C
  • Excellent electrical and mechanical properties offer a moisture seal and