Perfect-it microfibre wipes, 2022 series, Scotch-Brite™
Leverandør: 3M
2022
MMMA2022EA 0 NOK
MMMA2022
Perfect-it microfibre wipes, 2022 series, Scotch-Brite™
Papirservietter Rengjøringskluter for gjenbruk Kluter til generell bruk
The 2022 series Scotch-Brite™ Perfect-it microfibre wipes are premium microfibre polishing cloths. They are ideal for cleaning and polishing a wide range of surfaces such as stainless steel, chrome, glass, enamel, plastic, and lacquered furniture.
  • Microfibre wipes ideal for cleaning and polishing any surface
  • Provides a high-quality, lint-free finish without streaks or marks
Clean and polish any surface with Scotch-Brite™ Perfect-it Microfibre Wipes, 2022 series. These premium microfibre polishing cloths are ideal for cleaning and polishing.