Safety overspectacles, 2800 series
Leverandør: 3M
2801
MMMA2801EA 240 NOK
MMMA2801
Safety overspectacles, 2800 series
Vernebriller
3M™ Safety Overspectacles 2800 Series are over-eyewear safety spectacles featuring inclinable lenses and four low-profile temple lengths. The strong polycarbonate lens features an anti-scratch coating.
  • Over The Glass (OTG) range designed to fit over most prescription glasses
  • Excellent coverage and field of vision, combined with a high level of protection against impacts
  • Adjustable ratchet temples to adjust the lens for facial features and eyeglass position
  • Four adjustable low-profile temple lengths to ensure an optimal fit
For eye protection whichMore Product Information