Box sealing tape, 3444, Scotch®
Leverandør: 3M
344450500
MMMA344450500EA 1975 NOK
MMMA344450500
Box sealing tape, 3444, Scotch®
Tape
The 3M 3444 paper packaging adhesive tape is applied to dry and dust-free cardboard surfaces. It is characterised by its good holding power and long-term adhesion to paperboard. This makes the adhesive tape a very good alternative to classic film tapes. The special feature is that the adhesive tape retains its adhesion at all temperatures to which a package is usually exposed. This also applies to the deep-freeze area.
  • Adhesive: Natural rubber
  • Material type: Paper