Taper, blyfolie, 420, 3M™
Leverandør: 3M
4202533
MMMA4202533EA 3900 NOK
MMMA4202533
Taper, blyfolie, 420, 3M™
Tape
This lead foil tape has a lead foil backing with a green rubber adhesive and a clear, easy-release film liner.
  • Good conformability
  • Resists flame, moisture, weather and UV degradation
  • Electrically and thermally conductive
  • Strong acid and base resistance
  • Acts as a radiation barrier
  • Can be die-cut into specific shapes
The tape is used as a maskant in electroplating and as a moisture and radiation barrier in certain applications. The lead foil backing provides good conformability and has electrically- and thermally-conductive properties.
Length: 33,0 m
Colour: Silver