Boot covers, 440
Leverandør: 3M
440
MMMA440EA 6125 NOK
MMMA440
Boot covers, 440
Engangsplagg
These boot covers are made of combined SMS laminate and provide protection agaoinst non hazardous liquids and dust.
  • Anti-static treated
  • With ties for an excellent fit
  • Serged seams
Length: 406 mm.
Sertifiseringer: Cat. I.