Extreme sealing tape 4411N
Leverandør: 3M
4411N
MMMA4411NEA 985 NOK
MMMA4411N
Extreme sealing tape 4411N
Tape
This is a neutral, 1 mm thick, highly conformable, single-side acrylic tape with a high performance ionomer backing. It is specifically designed to provide professional results under outdoor exposure to direct sunlight, as well as temperature and humidity extremes, while creating an effective seal over an existing joint, seam, or penetration. It is part of the industrial professional range, which is supplied in a blister pack with multi-language descriptions.
  • Very good resistance to many common industrial solvents as well as elevated temperature exposure