Overalls, 4500
Leverandør: 3M
4500WS 4500WM 4500WXL 4500W2XL 4500W3XL 4500W4XL 4500WL
MMMA4500WSEA 2425 NOK
MMMA4500WS MMMA4500WM MMMA4500WXL MMMA4500W2XL MMMA4500W3XL MMMA4500W4XL MMMA4500WL
Overalls, 4500
Engangsplagg
This simple overall made of durable PP fabric is suitable for non hazardous applications where hygiene and cleanliness are important.
  • Excellent economic solution to keep the wearer clean or dry
  • Breathable and light duty protection
  • Two-way zip with storm flap
  • Elasticated 2-panel hood, cuffs, ankles and waist
Typical applications include: Domestic DIY use, light duty industrial cleaning and maintenance applications.
Weight: 45 g/m²
Sertifiseringer: Cat. I
Bestill nå
SPESIFIKASJONER
Modell 4500
Environmentally Preferable
Stoff PP
Kjønn
Antrekksapplikasjoner General Purpose,Laboratory,Industrial
Weight 45 g/m²