Glass cloth tape, 5453
Leverandør: 3M
54537533
MMMA54537533EA 67900 NOK
MMMA54537533
Glass cloth tape, 5453
Tape
Premium performance 3M™ PTFE glass cloth tape 5453 has a brown woven glass cloth backing impregnated with PTFE (polytetrafluoroethylene). This tape provides a slick surface on process machinery and offers an extremely low coefficient of friction which, in combination with a pressure sensitive silicone adhesive, creates a non-stick surface that performs at elevated temperatures.
  • Glass cloth reinforced backing provides greater tensile strength and resistance to tears and punctures than typical PTFE film backed tapes
  • Silicone adhesive offers high temperature