Face shield, polycarbonate
Leverandør: 3M
5E-11
MMMA5E-11EA 880 NOK
MMMA5E-11
Face shield, polycarbonate
Ansiktsskjermer
Face Shield 5E-11 is an optical class 1, shade 5, green tinted, curved polycarbonate faceshield with anti-fog and anti-scratch coating for protection against high speed particles, UV, and intense visible light. It is compatible with the V5 Faceshield System for mounting on 3M Hard Hat Series G3000 or G22.
  • Provides protection against high speed particles and secondary welding light
  • Designed to fit 3M Hard Hat Series G3000 or G22 with 3M V5 Faceshield Mounting system
  • Snap-in feature makes it quick and easy to connect to the face shield holder
  • Medium energy