Face Shields, V5 series
Leverandør: 3M
5F-11
MMMA5F-11EA 425 NOK
MMMA5F-11
Face Shields, V5 series
Ansiktsskjermer
Optical class 1, clear curved polycarbonate faceshield with anti-fog and anti-scratch coating. Compatible with 3M™ V5 Faceshield System for mounting on 3M™ Hard Hat Series G3000 or G22.
  • Provides protection against secondary welding light
  • Anti-fog and anti-scratch coated
  • Snap in feature makes it quick and easy to connect
  • Medium energy impact, Grade B
  • Optical class 1 visor, low energy impact
Get protection against impacts and secondary welding light with 3M™ Faceshield, 5F-11. It provides impact protection B against medium speed particles (0.86 g steelMore Product Information