Gass-, damp- og partikkelfiltre, 6000-serien
Leverandør: 3M
6051 6054 6055 6057 6059 6075 6098 6099 6096 6035 6035UK 6054UK 6055UK 6057UK 6059UK 6051UK 6075UK 6096UK 6098UK 6099UK 6095 6092
111-0051EA 1450 NOK
111-0051 111-0052 111-0053 111-0054 111-0058 111-8930 111-8932 111-0059 111-8931 111-0244 111-2188 MMMA6054UK 111-2191 111-2192 111-2193 MMMA6051UK 111-2194 111-2195 MMMA6098UK 111-2196 MMMA6095 MMMA6092
Gass-, damp- og partikkelfiltre, 6000-serien
Respiratorer Flerbruksmasker Filter for vernemasker
The 3M 6000 Series Gas, Vapor, and Particulate Combination Filters have a bayonet-style connection that fits any half and full-face 3M reusable masks. These lightweight filters offer well-balanced breathing protection for hazardous environments, and are designed to optimise the field of vision.
  • Protects against range of vapors, gases, and particulates
  • Combines protection with comfort and balance
  • Designed to optimize the field of vision
  • Lightweight with low breathing resistance
  • Bayonet-style connection fits a wide range of 3M reusable masks
  • Twist and click