3M™ PELTOR™ X1B Earmuffs Neckband
Leverandør: 3M
7100123139
MMMA7100123139EA 0 NOK
MMMA7100123139
3M™ PELTOR™ X1B Earmuffs Neckband
Hørselvern Hørselvern med lydkontroll
3M™ PELTOR™ X1 earmuffs are our lightweight hearing protectors with comfortable, soft ear cushions. They feature an electrically insulated, twin headband that decreases heat build-up on top of the head, and offer noise reduction up to 27 dB.
  • Neckband model for those wearers who prefer to wear an over the head band or wear multiple head and face PPE where a backband is more compatible
  • Colour-coding makes it easy to identify the level of protection being worn
  • Slim cups offer improved compatibility with other safety equipment
  • Wide soft ear cushions help