Tegaderm™ Transparent Film Roll
Leverandør: 3M
7100221528
MMMA7100221528EA 0 NOK
MMMA7100221528
Tegaderm™ Transparent Film Roll
Førstehjelpsutstyr Bandasjer
Non-sterile version of Tegaderm™ Transparent film roll provides all the benefits of surgical tape while providing a waterproof, bacterial and viral barrier.
  • The conforming film moves with the skin for greater patient comfort
  • Breathable foil allows oxygen and steam to pass through
  • It can be cut and adjusted to the required length and shape
  • Easy to apply with gloves on
  • Waterproof
  • Industrial category: Safety
In vitro testing shows that the transparent film provides a viral barrier from viruses 27 nm in diameter or larger while the dressing remains intactMore Product Information