Polyester Film Tapes, 850
Leverandør: 3M
850R50 850T19 850T2566 850S5166
MMMA850R50EA 18375 NOK
MMMA850R50 MMMA850T19 MMMA850T2566 MMMA850S5166
Polyester Film Tapes, 850
Tape
3M™ polyester tape 850 is designed for splicing applications requiring quick stick and an aggressive, permanent hold. Used in a variety of applications, including web processing, label protection, and marking.
  • Aggressive acrylic adhesive provides quick stick and excellent long-term holding in splicing applications
  • Thin, high-strength polyester film meets low calliper requirements and will not interfere with most machinery processes
  • Polyester backing resists tearing and protects surfaces from abrasion and rough handling
  • Temperature resistance enables tape