Polyester tape 8992
Leverandør: 3M
8992
MMMA8992EA 690 NOK
MMMA8992
Polyester tape 8992
Tape
3M™ Polyester Tape 8992 is a general-purpose tape that excels in high temperature masking operations such as powder coat painting. Coated with a silicone adhesive, this tape bonds to a variety of difficult surfaces. It is chemical-resistant and performs under a wide range of temperatures and environments.
  • Works in demanding conditions and high temperatures
  • Polyester film offers excellent thermal and chemical resistance while maintaining flexibility
  • Thicker version of 3M™ Polyester Tape 8991
  • Silicone adhesive provides clean removal from a variety of