Tapes, double coated, 9191
Leverandør: 3M
9191
MMMA9191EA 12575 NOK
MMMA9191
Tapes, double coated, 9191
Tape
3M™ double coated tape 9191 is a general-purpose double-coated tape. It features a high tack, high initial adhesion rubber pressure sensitive adhesive, a non-woven carrier, and an easy to remove siliconised paper liner.
  • General-purpose, double sided tissue tape
  • High tack, high initial adhesion
  • Pressure sensitive adhesive
  • Carpet tape
  • Cloth back material
With excellent balance between high tack/initial adhesion and good room temperature shear resistance as well as good removability, 9191 tape has been developed as a carpet tape. It is, however,More Product Information