Scotch® 926 ATG Adhesive Transfer Tape
Leverandør: 3M
926633
MMMA926633EA 490 NOK
MMMA926633
Scotch® 926 ATG Adhesive Transfer Tape
Tape
Scotch® ATG Adhesive transfer tape 926 is a safe alternative to hot melt adhesives. This tape features high initial adhesion and good holding power to a variety of materials including most plastics. This transfer tape is an acrylic, reverse wound, pressure sensitive adhesive on a densified kraft paper liner that can be used with Scotch® ATG Adhesive applicators.
  • High performance adhesive transfer film tape works a wide variety of substrates and materials
  • Offers excellent solvent resistance and can be used in high temperature applications
  • Bonds on contact,