Removable repositionable double coated tape, 9425HT
Leverandør: 3M
9425HT
MMMA9425HTEA 58700 NOK
MMMA9425HT
Removable repositionable double coated tape, 9425HT
Tape
This 152 µm double coated tape with high tack acrylic adhesive 420 on one side and medium tack acrylic adhesive 1050 on the other side, is supplied on a 58 lbs. polycoated kraft paper liner with a PET carrier.
  • 1,4 mil PET carrier coated with 2,2 mil 420 adhesive on face side and 2,2 mil 1050 adhesive on back side
  • Suitable for high temperature applications
  • Position and reposition lightweight materials
  • Works on a variety of substrates or materials for temporary mounting of lightweight materials
Temporarily fix or mount lightweight materials or createMore Product Information