Adhesive Transfer Tape, 966
Leverandør: 3M
966300
MMMA966300EA 13175 NOK
MMMA966300
Adhesive Transfer Tape, 966
Tape
Adhesive transfer tape 966 leverages the power of 3M™ high temperature acrylic adhesive 100 to produce an excellent high temperature, high strength, and chemical-resistant bond suitable to the demands of aerospace, automotive, and electronics markets. It provides low outgassing properties necessary to deliver optimal performance in demanding applications.
  • Transparent, double sided, 50 µm adhesive transfer tape
  • High temperature acrylic adhesive for a long-lasting bond
  • Excellent solvent and UV resistance
  • High peel strength compared to other acrylic adhesives