Tape, Double Coated, 9731
Leverandør: 3M
9731
MMMA9731EA 117400 NOK
MMMA9731
Tape, Double Coated, 9731
Tape
The tape is developed with a high-performance acrylic adhesive on the face side and a silicone adhesive on the liner side of a 1 mil clear PET carrier.
  • Double linered - Adhesive 350
  • Ideal for attaching silicone rubber elastomers and foam
Features acrylic and silicone adhesives for specialised applications such as silicone gasketing. A thin polyester carrier provides dimensional stability and improved handling with ease of die cutting and lamination compared to adhesive transfer tapes.
It is excellent for demanding applications where siliconeMore Product Information