Adhesive applicator, ATG 700, Scotch®
Leverandør: 3M
ATG700EU
MMMAATG700EUEA 6200 NOK
MMMAATG700EU
Adhesive applicator, ATG 700, Scotch®
Dispensere og stempler for flytende lim
Scotch® ATG adhesive applicators apply adhesive transfer tape in a quick, easy, single-handed operation. These lightweight applicators are ideal for use on small and medium assembly jobs, as well as light and heavy-duty industrial applications.
  • Applicator accommodates the use of ¾" and ½" width adhesive rolls
  • Controlled application with the squeeze of a trigger
  • Applies adhesive while removing and rewinding liner back into the applicator to eliminate clean-up
  • Transparent cover allows quick and easy inspection to show remaining tape
  • Universal for varoius