Banded earplugs, E-A-Rcaps™
Leverandør: 3M
EC01000S
MMMAEC01000SEA 0 NOK
MMMAEC01000S
Banded earplugs, E-A-Rcaps™
Ørepropper Ørepropper med bøyle
The E-A-Rcaps™ banded hearing protector is fitted with semi-aural eartips designed to seal the entrance of the ear canal to help reduce exposure to hazardous levels of noise.
  • Reduces noise levels up to 23 dB
  • Flexible and durable band can be washed and reused multiple times
  • Soft foam pod shaped caps to block the ear canal
  • Ideal for intermittent wear, the band can be easily stored around the neck when not in use
  • Compatible with the 3M™ E-A-Rfit™ Dual-Ear validation system
Get one of the lightest banded hearing protectors on the market with 3M™ E-A-Rcaps™More Product Information