Manual dispenser and applicator, Scotch-Weld™ EPX™
Leverandør: 3M
EPX400H
MMMAEPX400HEA 15500 NOK
MMMAEPX400H
Manual dispenser and applicator, Scotch-Weld™ EPX™
Klebemidler
Manual applicator for dispensing EPX adhesive in 38 and 50 ml cartridges supplied with two plungers for 1:1/2:1 and 10:1 adhesive.
  • Dispenses two-component 3M™ Scotch-Weld™ structural adhesive
  • Offers take-it-to-the-job convenience without the need for air hoses or electrical cords
  • Ensures correct mix ratio during dispensing; no need to weigh components
  • Minimises waste and mess associated with hand mixing
  • Reduce plastic waste
Dispense 3M™ Scotch-Weld™ adhesive in duo-pack cartridges with 3M™ Scotch-Weld™ EPX manual applicator. Our applicator mixes andMore Product Information