Scotch-Weld™ EPXG50 Manual Applicator
Leverandør: 3M
EPXG50
MMMAEPXG50EA 2350 NOK
MMMAEPXG50
Scotch-Weld™ EPXG50 Manual Applicator
Dispensere og stempler for flytende lim
The 3M™ Scotch-Weld™ EPX glue guns make it clean and easy to apply 3M™ Scotch-Weld™ two-component structural adhesives with minimal waste.
  • Ensure accurate measurement
  • Complete mixing and precise material placement
  • For 38/45/48,5 and 50 ml cartridges
Manual hand movement drives both pistons directly into the Duo-Pak cartridge, applying equal pressure and feeding both components of the adhesive directly into the mixing nozzle (sold separately). These handheld applicators are convenient to use on the job-no air hose or electrical cord is required.