Elevated Temperature Front Helmet Cover
Leverandør: 3M
FC1-AL
MMMAFC1-ALEA 2150 NOK
MMMAFC1-AL
Elevated Temperature Front Helmet Cover
Hjelmer Sikkerhethjelm
The elevated temperature front cover is a silver aluminium front cover designed to fit on to 3M™ hard hats.
  • Provide additional protection from sparks and metal splatter
  • Streamlined design prevents interference with other equipment during use
  • Fabric parts can be easily replaced when needed
Get extra protection from sparks and metal splatter on the front of a hard hat with the elevated temperature front cover. This silver aluminium hard hat cover has a streamlined design that helps prevent interference with other equipment during use. It fits on to 3M™More Product Information