Hard hat replacement suspension system, G2C
Leverandør: 3M
G2C
MMMAG2CEA 260 NOK
MMMAG2C
Hard hat replacement suspension system, G2C
Hjelmer Sikkerhethjelm
3M™ pinlock suspension, G2C, is the replacement pinlock suspension cradle and plastic sweatband for all 3M™ hard hats (except the X5000 series).
  • Replacement pinlock suspension cradle with plastic sweatband
  • Pinlock adjustment system provides simple and secure size adjustment for head sizes from 540 to 620 mm
  • Reliable head and face protection products meet regulated performance and safety requirements
Replace the pinlock suspension and plastic sweatband on hard hats with 3M™ pinlock suspension, G2C. This replacement pinlock suspension cradle snapsMore Product Information