Hard Hat Replacement Suspension Systems, G/H
Leverandør: 3M
G2N
MMMAG2NEA 395 NOK
MMMAG2N
Hard Hat Replacement Suspension Systems, G/H
Hjelmer Sikkerhethjelm
3M™ Ratchet suspension, G2N is a replacement ratchet suspension cradle and plastic sweatband for all 3M™ Hard Hats, except the X5000 series.
  • Replacement ratchet suspension cradle with plastic sweatband
  • Ratchet adjustment system provides simple and secure size adjustment
  • Suitable for use with all 3M™ hard hats
Replace the ratchet suspension and plastic sweatband on a hard hat with 3M™ Ratchet Suspension, G2N. This replacement ratchet suspension cradle is designed for all 3M™ Hard Hats, except the X5000 series. It snaps easily into place to provide allMore Product Information