Safety goggles, GoggleGear™ 6000 series
Leverandør: 3M
GG6001SGAF-BLK GG6001SGAF-GRN GG6001SGAF-RED GG6001NSGAFBLU
MMMAGG6001SGAF-BLKEA 620 NOK
MMMAGG6001SGAF-BLK MMMAGG6001SGAF-GRN MMMAGG6001SGAF-RED MMMAGG6001NSGAFBLU
Safety goggles, GoggleGear™ 6000 series
Vernebriller /Goggles
GoggleGear™ safety goggles 6000 series features a large profile with indirect ventilation for air circulation with Scotchgard™ anti-fog and anti-scratch coating.
  • Large profile safety goggles provide enough space to fit over most prescription glasses
  • Indirect airflow to help against fogging
  • Premium spherical curved lens design for optimal horizontal and vertical field of vision
  • Scotchgard™ anti-fog coating provides superior anti-fog and anti-scratch properties
  • Pivoting ratchet for convenient personal adjustment
  • 2C-1.2 3M 1 BT K N lens marking
  • Wide flexible