Double coated tape, GPT-020F
Leverandør: 3M
GPT-020F1540
MMMAGPT-020F1540EA 23325 NOK
MMMAGPT-020F1540
Double coated tape, GPT-020F
Tape
Designed for versatility, 3M™ double coated tape GPT-020F is a transparent, 200 µm thick, modified solventless acrylic adhesive with a white printed PP filmic liner. It combines a high level of peel and shear performance, enabling high adhesion to a wide variety of substrates.
  • Providing the capability to bond to a wide variety of substrates, from high to low surface
  • High initial tack and humidity resistance
  • Easy handling and converting due to filmic liner delivery
  • 200 µm thick
This tape offers high initial tack and humidity resistance, providing easyMore Product Information