Earmuffs, Peltor™ Optime III™
Leverandør: 3M
H540AV
MMMAH540AVEA 985 NOK
MMMAH540AV
Earmuffs, Peltor™ Optime III™
Hørselvern Hørselvern med lydkontroll
The earmuffs are designed specifically for heavy noise and combine effective protection with the ultimate in comfort. The model H540AV with headband is in Hi-VIZ design for high visibility. The advantage like conversations and warning signals are still easy to understand. An acoustic connection between the capsule interior and the space between the shells also achieves optimal low-frequency insulation.
  • Double capsule construction for highest insulation performance
  • Filled with a unique combination of liquid and foam
  • Perfect adaptation to the head shape
  • Very