Earmuffs, Peltor™, WorkTunes™
Leverandør: 3M
HRXS220A
MMMAHRXS220AEA 3700 NOK
MMMAHRXS220A
Earmuffs, Peltor™, WorkTunes™
Hørselvern Hørselvern med radio
The headset helps to protect from potentially harmful workplace noise while at the same time the wearer can listen to the built in AM/FM radio.
  • Digital station search and voice guided menu system with announced frequencies
  • 3,5 mm listen only stereo input (limited to 82 dB) for connection to external devices (e.g. mobile phone, radio, two-way radio and iPod) and a voice guided menu system to avoid you having to take the headset off to change or check settings
  • Auto power off turns headset off after 4 hours of non-use, to save energy and a low-battery warning