Adhesive remover, spray, Scotch-Weld™
Leverandør: 3M
IR500
MMMAIR500EA 1075 NOK
MMMAIR500
Adhesive remover, spray, Scotch-Weld™
Rengjøringsløsninger Detergenter og overflatevasking
Powerful citrus-based adhesive remover spray cleaner effectively removes adhesive residue, grease and tar.
  • Clean and degrease metal parts, motors, tools, printing presses and concrete
  • Clean annoying residue from sticky labels
  • Aerosol spray can allows quick and easy application
  • Pleasant citrus smell