Earmuffs, Peltor™ Alert™
Leverandør: 3M
M2RX7A
MMMAM2RX7AEA 6400 NOK
MMMAM2RX7A
Earmuffs, Peltor™ Alert™
Hørselvern Hørselvern med radio
This high-attenuating over-the-head hearing protection headset incorporates an electronic level dependent AM/FM radio receiver with approximately 70 hours of operating time on two standard AA batteries.
  • Level dependent function attenuates loud noises while amplifying weak ones
  • Conveniently powered by two standard AA batteries
  • Obtain crisp AM/FM radio signals on digital 2G circuit
  • Easty to handle volume control knobs for radio and level dependent function
  • Wide and soft earcups offer comfort for long wear and optimum seal
Ideal for use in the followingMore Product Information