Powered air respirator headtop collar
Leverandør: 3M
M444
MMMAM444EA 1475 NOK
MMMAM444
Powered air respirator headtop collar
Hjelmer Sikkerhethjelm
Replacement inner collar for use with the Versaflo™ M-400 Headtop. The inner collar zips into the shroud and provides a comfortable respiratory seal.
  • Replacement inner collar
  • Provides a comfortable respiratory seal
  • Made from polypropylene coated knitted polyamid
Replacement inner collar is designed for use with the 3M™Versaflo™ M-400 Series Helmets.