Earmuffs, WS™ Alert™ XPI headset, Peltor™
Leverandør: 3M
MRX21A3WS6
MMMAMRX21A3WS6EA 14725 NOK
MMMAMRX21A3WS6
Earmuffs, WS™ Alert™ XPI headset, Peltor™
Hørselvern Hørselvern med radio
The Peltor™ WS™ Alert™ XPI Headset is a is high-attenuating hearing protector that can connect up to two Bluetooth® enabled devices, such as a mobile phone and two-way radio. This headset also has a noise-cancelling boom microphone, level-dependent function for ambient listening, push-to-listen feature and built-in FM radio.
  • Protects against hazardous noise
  • Enables hands-free calls in noisy environments
  • iOS and Android compatibility with 3M™ connected Equipment app
  • Stand-alone headset with headband
  • Bluetooth® MultiPoint technology connects up to two