Earmuffs, Peltor™ WS™ ProTac XPI headsets
Leverandør: 3M
MT15H7AWS6-111 MT15H7AWS6 MT15H7P3EWS6
MMMAMT15H7AWS6-111EA 19400 NOK
MMMAMT15H7AWS6-111 MMMAMT15H7AWS6 MMMAMT15H7P3EWS6
Earmuffs, Peltor™ WS™ ProTac XPI headsets
Hørselvern Hørselvern med radio
Peltor™ WS™ ProTac XPI is a range of level-dependent Bluetooth® headsets for effective hearing protection without compromising communication. They come in a high visibility yellow colour, with a headband, neckband, or helmet attachment, and are also available with FLX2 accessory connectors.
  • Level-dependent for ambient listening
  • Bluetooth® connectivity for seamless connection
  • Bluetooth® multiPoint for simultaneous connection of up to two devices
  • IP68 classified noise-cancelling boom microphone for easy inter-user communication
  • High visibility yellow for