Earmuffs, WS™ LiteCom Plus headsets, Peltor™
Leverandør: 3M
MT73H7P3E4410E MT73H7A4310WEU MT73H7B4310WEU MT73H7P3E4310E
MMMAMT73H7P3E4410EEA 31325 NOK
MMMAMT73H7P3E4410E MMMAMT73H7A4310WEU MMMAMT73H7B4310WEU MMMAMT73H7P3E4310E
Earmuffs, WS™ LiteCom Plus headsets, Peltor™
Hørselvern Hørselvern med radio
The Peltor™ WS™ LiteCom Plus Headset is available in multiple mounting styles, and features an integrated pre-programmed analog two-way radio, Bluetooth® multipoint connectivity, noise-cancelling microphone, and a level-dependent function for ambient listening.
  • Hearing protector with built-in analogue two-way radio
  • Level-dependent technology improves situational awareness and communication
  • Bluetooth® multipoint connectivity allows seamless connection of mobile devices
  • Push-to-listen quick activation of level dependent function
  • Noise cancelling waterproof