Integrated suspension hoods, Versaflo™ S-400 Series
Leverandør: 3M
S-433L-5
MMMAS-433L-5EA 10225 NOK
MMMAS-433L-5
Integrated suspension hoods, Versaflo™ S-400 Series
Respiratorer Vernemaskesystem
3M™ Versaflo™ S-400 series integrated suspension hood is a head cover with integrated head suspension which provides head, face, neck, and shoulder coverage. The hood can be used with 3M™ powered air respirators and supplied air respirator systems in a wide variety of applications.
  • Protects face and eyes from liquid splash and low energy flying particles and dust (Grade F)
  • For use with certain 3M™ Powered Air Purifying and Supplied Air Respirator Systems
  • Features the 3M™ Quick Release Swivel (QRS) connection
  • General-purpose two-layer polypropylene spunbond/