Reusable Respirator Spectacle Kit
Leverandør: 3M
SK102
MMMASK102EA 1950 NOK
MMMASK102
Reusable Respirator Spectacle Kit
Respiratorer Flerbruksmasker Heldekkende ansiktsmasker
Wear prescription lenses while using certain 3M™ Reusable Full Face Masks with the help of 3M™ Reusable Respirator Spectacle Kit. These spectacle frames attach to 3M™ Reusable Full Face Masks and can be adjusted.
  • Spectacle kit for 3M™ Reusable Full Face Masks
  • Permits wearer to use corrective lenses and to adjust them to a comfortable position
  • One each per case
  • Easy installation
  • For occupational use only
Get the correct level of comfort and protection from protective equipment, while using prescription spectacles, with the 3M™ Reusable RespiratorMore Product Information