Powered air respirator spark arrestor for Versaflo™ powered air turbo TR-300+, Versaflo™
Leverandør: 3M
TR362E
MMMATR362EEA 310 NOK
MMMATR362E
Powered air respirator spark arrestor for Versaflo™ powered air turbo TR-300+, Versaflo™
Respiratorer Vernemaskesystem
Versaflo™ Spark Arrestor TR-362 is an accessory designed to protect the filter and turbo in Versaflo™ Powered Air Turbo TR-300+.
  • Protect the filter and turbo unit from damage due to sparks or other hot materials
  • Installs inside the filter cover of Versaflo™ Powered Air Turbo TR-300+
The spark arrestor installs inside the filter cover of 3M™ Versaflo™ Powered Air Turbo TR-300+ to prevent hot materials from entering the turbo. It also acts as a pre-filter by capturing many of the larger particles that would otherwise be drawn into and clog the main filter.