Powered air respirator filter cover for Versaflo™ powered air turbo TR-300+, Versaflo™
Leverandør: 3M
TR371EPLUS
MMMATR371EPLUSEA 605 NOK
MMMATR371EPLUS
Powered air respirator filter cover for Versaflo™ powered air turbo TR-300+, Versaflo™
Respiratorer Vernemaskesystem
The Versaflo™ Filter Cover TR-371+ is a replacement filter cover for use with Versaflo™ Powered Air Turbo TR-300+.
  • Holds TR-3712E, TR-3802E, and TR-3822E filters in place on Versaflo™ Powered Air Turbo TR-300+
  • See filter label easily with quick view window
  • Easy to install replacement part
Hold filters securely in place on the turbo unit with 3M™ Versaflo™ Filter Cover, TR-371+. This filter cover is a replacement part which can be used with TR-3712E, TR-3802E, and TR-3822E filters. It’s easy to install and the quick view window enables users to easily see filter labels.