Earplugs, E-A-R™ Ultrafit™
Leverandør: 3M
UF01000S
MMMAUF01000SEA 0 NOK
MMMAUF01000S
Earplugs, E-A-R™ Ultrafit™
Ørepropper Flergangsørepropper
The 3M™ E-A-R™ UltraFit™ Earplugs are reusable and designed for insertion into the ear canal to help reduce exposure to hazardous levels of noise up to 32 dB.
  • Patented tri-flange design in pre-moulded material is a fit for most ear canals
  • Firm, long stem for easy insertion and removal
  • Made from soft and durable material
  • Separate cord can be removed and reinserted for optional uncorded or corded wear
  • Compatible with the 3M™ E-A-Rfit™ Dual-Ear Validation System
Get comfortable, durable and reusable hearing protection with 3M™ E-A-R™ UltraFit™ Earplugs.More Product Information